missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic acid, mono-2-ethyl-5-hydroxyhexyl ester 100 μg/mL in Methyl-tert-butyl ether, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Mono(2-ethyl-5-hydroxyhexyl) Phthalate (Mixture of Diastereomers) |
| CAS | 40321-99-1 |
| Molecular Formula | C16 H22 O5 |
| Quantity | 1 mL |
| Synonym | 1,2-Benzenedicarboxylic acid, mono(2-ethyl-5-hydroxyhexyl) ester (9CI), Mono-2-ethyl-5-hydroxyhexyl phthalate |
| InChI Formula | InChI=1S/C16H22O5/c1-3-12(9-8-11(2)17)10-21-16(20)14-7-5-4-6-13(14)15(18)19/h4-7,11-12,17H,3,8-10H2,1-2H3,(H,18,19) |
| SMILES | CCC(CCC(C)O)COC(=O)c1ccccc1C(=O)O |
| IUPAC Name | 2-(2-ethyl-5-hydroxyhexoxy)carbonylbenzoic acid |
| Molecular Weight (g/mol) | 294.34 |
| Formula Weight | 294.1467 g/mol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction