Learn More
Phthalic acid, bis-n-undecyl ester 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis undecyl ester |
| CAS | 3648-20-2 |
| Molecular Formula | C30 H50 O4 |
| Quantity | 1 mL |
| Synonym | Phthalic Acid, Bis-n-undecyl Ester, 1,2-Benzenedicarboxylic acid, diundecyl ester (9CI), Phthalic acid, diundecyl ester (7CI,8CI), Undecyl alcohol, phthalate (2:1) (8CI), Undecyl alcohol, phthalate (8CI), Bis-diundecylphthalate, Bisoflex DUP, DUP, Di-n-undecyl phthalate, Diplast L 11, Diundecyl phthalate, Jayflex L 11P, Palatinol 11, Sansocizer DUP |
| InChI Formula | InChI=1S/C30H50O4/c1-3-5-7-9-11-13-15-17-21-25-33-29(31)27-23-19-20-24-28(27)30(32)34-26-22-18-16-14-12-10-8-6-4-2/h19-20,23-24H,3-18,21-22,25-26H2,1-2H3 |
| SMILES | CCCCCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCCCCC |
| IUPAC Name | diundecyl benzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 474.72 |
| Formula Weight | 474.3709 g/mol |