missing translation for 'onlineSavingsMsg'
Learn More

Phthaldialdehyde, 98%

Product Code. p-7021103
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460883 10 g 10g
11470883 50 g 50g
11480883 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460883 Supplier Thermo Scientific Alfa Aesar Supplier No. A13299.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 643-79-8 | C8H6O2 | 134.13 g/mol

Phthaldialdehyde is used as a fluorescent reagent for checking the assay of amines present in proteins, peptides by capillary electrophoresis and chromatography. It is also used for cholesterol assay in the picomole range as well as for fluorometric determination of histamine, histidine and other amino acids. Poly(phthalaldehyde) is obtained by the polymerization reaction which is used to make a photoresist. The nitrogen by o-phthaldialdehyde assay (NOPA) is a method used in winemaking process to measure the completion of fermentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthaldialdehyde is used as a fluorescent reagent for checking the assay of amines present in proteins, peptides by capillary electrophoresis and chromatography. It is also used for cholesterol assay in the picomole range as well as for fluorometric determination of histamine, histidine and other amino acids. Poly(phthalaldehyde) is obtained by the polymerization reaction which is used to make a photoresist. The nitrogen by o-phthaldialdehyde assay (NOPA) is a method used in winemaking process to measure the completion of fermentation.

Solubility
Soluble in acetic acid and methanol. Slightly soluble in water.

Notes
Air, light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, amines and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 643-79-8
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.13
MDL Number MFCD00003335
InChI Key ZWLUXSQADUDCSB-UHFFFAOYSA-N
Synonym o-phthalaldehyde, o-phthaldialdehyde, benzene-1,2-dicarboxaldehyde, 1,2-benzenedicarboxaldehyde, phthaldialdehyde, phthalic aldehyde, phthalic dialdehyde, phthalyldicarboxaldehyde, ortho-phthalaldehyde, o-phthaldehyde
PubChem CID 4807
ChEBI CHEBI:70851
IUPAC Name phthalaldehyde
SMILES O=CC1=CC=CC=C1C=O

Specifications

Melting Point 53°C to 57°C
Flash Point 132°C (269°F)
Assay Percent Range 98%
Quantity 10 g
UN Number UN2923
Beilstein 878317
Sensitivity Air Sensitive
Solubility Information Soluble in acetic acid and methanol. Slightly soluble in water.
Formula Weight 134.14
Percent Purity 98%
Assay 98%
Chemical Name or Material o-Phthaldialdehyde
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phthalaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.