Learn More
Phthalan, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 496-14-0 |
| Molecular Formula | C8H8O |
| MDL Number | MFCD00005932 |
| Quantity | 100 g |
| SMILES | C12=C(COC2)C=CC=C1 |
| Molecular Weight (g/mol) | 120.15 |
| Percent Purity | 97% |
| Chemical Name or Material | Phthalan |