missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phoxim-d5 (phenyl-d5) (mixture of isomers), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Phoxim D5 (phenyl D5) |
| Molecular Formula | C12 D5 H10 N2 O3 P S |
| Quantity | 0.01 g |
| Synonym | Phoxim D5 (phenyl D5), Phoxim-d5, Phoxim-D5 (phenyl-D5), O,O-Diethyl O-(α-cyanobenzylideneamino-d5) phosphorothioate, Phenyl-d5-glyoxylonitrile oxime O,O-diethyl phosphorothioate, O-[[Cyano(phenyl-d5)methylene]azanyl]phosphorothioic acid O,O-diethyl ester, 4-Ethoxy-7-(phenyl-d5)-3,5-doxa-6-aza-4-phosphaoct-6-ene-8-nitrile 4-sulfide |
| InChI Formula | InChI=1 S/C12H15N2O3PS/c1-3-15-18(19,16-4-2)17-14-12(10-13)11-8-6-5-7-9-11/h5-9 H,3-4H2,1-2H3/i5D,6 D,7 D,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=NOP(=S)(OCC)OCC)C#N)c([2 H])c1[2 H] |
| IUPAC Name | 2,3,4,5,6-pentadeuterio-N-diethoxyphosphinothioyloxybenzenecarboximidoyl cyanide |
| Molecular Weight (g/mol) | 303.329 |
| Formula Weight | 303.085 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction