Learn More
Brady™ Photoluminescent Self-adhesive Dots
Product Code. 11766915 Shop All Brady Products
Change view
Click to view available options
Diameter (Metric):
16 mm
25 mm
Unit Size:
Pack of 15
Pack of 40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11766915 25 mm Pack of 15
11761316 16 mm Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11766915 Supplier Brady™ Supplier No. 263383
  • ONLINE EXCLUSIVE PRICE
    8.85€
    8.64€ / Pack of 15
    Save 0.21€
    2% Off
Estimated Shipment: 09-12-2026
Use these Photoluminescent Self-adhesive Dots to mark anything that is important to locate, especially in complete darkness.
  • Use these photoluminescent dots to mark anything that is important to locate, especially in complete darkness.
Specifications
Description Photoluminescent individual dots (Self-adhesive)
Adhesive Type Self-adhesive
Coating None
Color Photoluminescent
Display Blank
Finish Matte
Format Label
Material Polyester
Printable Sides Single-side Printable
Print Technology None
Product Type Photoluminescent Tape
Sterility Non-sterile
No. per Pack 15 Pieces/Card
Quantity 15 Pk.
Shape Round
Dimensions (L x W) 25 x 25 mm
Length (English) 2.5 in.
Width (English) 2.5 in.
Width (Metric) 63.5 mm
Diameter (English) 0.98 in.
Diameter (Metric) 25 mm
Weight (Metric) 0.01 kg
Alternate Part Number PHOTOLUM DISCS-DIA 2.5