missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Phosphorus Reagent Sets, For Colorimetric Water Testing

Product Code. 11798763
Click to view available options
Quantity:
1 Set
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 11798763

Brand: Hanna Instruments™ HI93758C50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Phosphorus Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.

These high quality reagents are designed to follow accepted test methods for colorimetric determination of various forms of phosphorus. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.

  • Reagent sets available for determinations of phosphorus, total phosphorus (high or low range), acid hydrolyzable phosphorus and reactive phosphorus (low or high range)
  • All sets except basic Phosphorus Reagents (HI93706-01) contain 50 ready-to-use vials as well reagent packets/dropper bottles
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability

Specifications

For Use With (Application) Total phosphorus determination
For Use With (Equipment) Compatible Hanna benchtop and portable photometers
Includes 50 ready-to-use vials of HI93758V-0AH, 1 bottle of HI93758C-0 solution, 50 packets of potassium persulfate reagent, and 50 packets of HI93758-0 phosphorus reagent
No. of Tests 50
Quantity 1 Set
Range 0 to 1.15mg/L phosphorus
Test Method EPA method 365.2 and 4500-P E ascorbic acid
Type Water Testing Kit
compliance-icons compliance-icons
Product Identifier
  • X50 TEST TOTAL P 0-3,5MG/L
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : potassium disulphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.