missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Phosphoramidon disodium salt
GREENER_CHOICE

Product Code. 18775516
Click to view available options
:
5mg
This item is not returnable. View return policy

Product Code. 18775516

Brand: Tocris Bioscience™ 6333/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Neutral endopeptidase and endothelin converting enzyme inhibitor

Phosphoramidon disodium salt is a neutral endopeptidase (neprilysin) inhibitor. Blocks degradation of amyloid β peptides and increases Aβ levels in rodents. Also blocks endothelin converting enzyme (ECE).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[N-[[(6-Deoxy-α-L-mannopyranosyl)oxy]hydroxyphosphinyl]-L-leucyl]-L-tryptophan disodium salt
CAS 164204-38-0
Assay Percent Range >95%
Quantity 5 mg
Molecular Formula C23H32N3Na2O10P
Formula Weight Observed MW: 587.47
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.