missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Phospho-GSK-3 alpha/beta (S21/S9) DuoSet IC ELISA

Product Code. p-4754907
Click to view available options
Format:
2 96-well Plates
5 96-well Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30021399

Brand: R&D Systems™ DYC26302

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

The Phospho-GSK-3 alpha/beta (S21/S9) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat GSK-3 alpha/beta.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Kit
Quantity 1 Kit
Species Human, Mouse, Rat
Conjugate Biotin
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Format 2 96-well Plates
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.