missing translation for 'onlineSavingsMsg'
Learn More

Chemetrics™ Phosphate CHEMets™ Refill

Product Code. 11771054
Click to view available options
Quantity:
30 Ampules
Unit Size:
Pack of 30
This item is not returnable. View return policy

Product Code. 11771054

Brand: Chemetrics™ R8515

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Analytes Phosphate, ortho
For Use With (Equipment) Refill kit for K-8515 or K-8530
No. per Pack 30
Quantity 30 Ampules
Test Method Vanadomolybdophosphoric Acid
Test Time 5 min.
compliance-icons compliance-icons
Product Identifier
  • Phosphate Vacu-vials and CHEMets refill
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P234-Keep only in original container.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to authorised chemicallandfill or if organic to high temperature incineration.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, ammonium molybdate, ammonium metavanadate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.