Learn More
Tocris Bioscience™ Phortress
Prodrug of the antitumor agent 5F 203
Brand: Tocris Bioscience™ 4995/10
This item is not returnable.
View return policy
Description
Phortress is a prodrug of the antitumor agent 5F 203, which acts via binding to aryl hydrocarbon receptors. Induces expression of CYP1A1 and generates adducts in the DNA of sensitive MCF7 and IGROV-1 cells.Specifications
| >98% | |
| 10 mg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.