Learn More
Tocris Bioscience™ Phortress
GREENER_CHOICE
Prodrug of the antitumor agent 5F 203
Brand:  Tocris Bioscience™ 4995/10
Encompass
Product Code. 15293347
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
Phortress is a prodrug of the antitumor agent 5F 203, which acts via binding to aryl hydrocarbon receptors. Induces expression of CYP1A1 and generates adducts in the DNA of sensitive MCF7 and IGROV-1 cells.
Specifications
Specifications
>98%
10 mg