Description
- Sporty safety spectacles with high wearer acceptance
- Robust polycarbonate lens with proven uvex supra-vision coating technology - permanently anti-fog and scratch-resistant
- Metal-free
- Comfort features X-tended eyeshield: Soft-seal frame technology wraps around the forehead and provides protection against particle penetration
- Ergonomically shaped side arms with soft ear-pieces ensure a comfortable and secure fit without creating pressure points
- Soft, flexible nose piece adapts to the wearer to deliver a non-slip, pressure-free fit
- Certified in accordance with EN 166 (personal eye protection) and EN 170 (UV filters) UV400 -100% protection against harmful UV radiation up to 400 nm
Specifications
Specifications
| Lens Coating | Anti-fog, Anti-scratch, Scratch-resistant |
| Lens Tint | Amber |
| Product Type | Safety Glasses |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | TPE |
| UV Protection | UV400 |
| Frame Type | Semi-rimless |