missing translation for 'onlineSavingsMsg'
Learn More

(Phenylthio)acetaldehyde Diethyl Acetal, TRC

Product Code. 30426717
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426717 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426717 Supplier Toronto Research Chemicals Supplier No. TRCP33707510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (Phenylthio)acetaldehyde Diethyl Acetal
CAS 66616-26-0
Molecular Formula C12H18O2S
Quantity 10 mg
Synonym (Phenylthio)acetaldehyde Diethyl Acetal, [(2,2-Diethoxyethyl)thio]benzene
InChI Formula InChI=1S/C12H18O2S/c1-3-13-12(14-4-2)10-15-11-8-6-5-7-9-11/h5-9,12H,3-4,10H2,1-2H3
SMILES CCOC(CSC1=CC=CC=C1)OCC
IUPAC Name (2,2-diethoxyethyl)(phenyl)sulfane
Molecular Weight (g/mol) 226.34
Formula Weight 226.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.