missing translation for 'onlineSavingsMsg'
Learn More

Phenylglyoxal monohydrate, 97%

Product Code. 11453434
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11453434

Brand: Thermo Scientific Alfa Aesar A10677.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1075-06-5 | C8H8O3 | 152.149 g/mol

Phenylglyoxal monohydrate was used to modify pig muscle carbonic anhydraseIII and as a specific reagent for arginine groups. It was also used to prepare pyrrolinone and furan derivatives and as as a chemiluminescent reagent for the determination of purines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylglyoxal monohydrate was used to modify pig muscle carbonic anhydraseIII and as a specific reagent for arginine groups. It was also used to prepare pyrrolinone and furan derivatives and as as a chemiluminescent reagent for the determination of purines.

Solubility
Partly miscible with water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1075-06-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00149499
InChI Key NBIBDIKAOBCFJN-UHFFFAOYSA-N
Synonym dihydroxyacetophenone, acetophenone, dihydroxy, dihydroxy 1-phenylethanone, 2,2-dihydroxy-1-phenyl-ethanone, ethanone, 2,2-dihydroxy-1-phenyl, acetophenone, 2,2-dihydroxy, alpha,alpha-dihydroxyacetophenone, dioxyacetophenone, alpha,alpha-dihydroxy-acetophenone
PubChem CID 99611
IUPAC Name 2,2-dihydroxy-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(O)O

Specifications

Melting Point 71°C to 77°C
Boiling Point 142°C (125 mmHg)
Quantity 25 g
Beilstein 1854721
Solubility Information Partly miscible with water.
Formula Weight 152.15 (134.12 Anhydrous)
Percent Purity 97%
Chemical Name or Material Phenylglyoxal monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.