Learn More
Phenyl beta-D-galactopyranoside, 99%
Product Code. 11481690
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11481690
Brand: Thermo Scientific Alfa Aesar H27028.06
Request Bulk or Custom Format
CAS: 2818-58-8 | C12H16O6 | 256.254 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2818-58-8
Molecular Formula C12H16O6
Molecular Weight (g/mol) 256.254
MDL Number MFCD00063258
InChI Key NEZJDVYDSZTRFS-YBXAARCKSA-N
Synonym phenylgalactoside, phenyl beta-d-galactopyranoside, phenyl b-d-galactoside, phenyl-b-d-galactopyranoside, phenyl b-d-galactopyranoside, phenyl-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-phenoxy-tetrahydropyran-3,4,5-triol, p-gal, phenylb-d-galactoside, phenyl, a-d-galactoside
PubChem CID 102336
ChEBI CHEBI:8098
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol
SMILES C1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O
Specifications
Melting Point 152°C to 156°C
Quantity 5 g
Beilstein 87518
Optical Rotation −42° (c=2 in Water)
Formula Weight 256.25
Percent Purity 99%
Chemical Name or Material Phenyl beta-D-galactopyranoside