Learn More
Phenyl-2,3,4,6-tetra-O-acetyl-Beta-D-galactopyranoside, TRC
Specifications
Specifications
| Chemical Name or Material | Phenyl-2,3,4,6-tetra-O-acetyl-beta-D-galactopyranoside |
| CAS | 2872-72-2 |
| Molecular Formula | C20H24O10 |
| Quantity | 10 g |
| Synonym | Phenyl Tetra-O-acetyl-beta-D-galactopyranoside, NSC 173175 |
| InChI Formula | InChI=1S/C20H24O10/c1-11(21)25-10-16-17(26-12(2)22)18(27-13(3)23)19(28-14(4)24)20(30-16)29-15-8-6-5-7-9-15/h5-9,16-20H,10H2,1-4H3/t16-,17+,18+,19-,20-/m1/s1 |
| SMILES | CC(=O)OC[C@H]1O[C@@H](Oc2ccccc2)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@H]1OC(=O)C |
| IUPAC Name | [(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-phenoxyoxan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 424.4 |
| Formula Weight | 424.1369 |