Learn More
Phenoxybenzamine Hydrochloride, TRC
Product Code. 30416754
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30416754 50 mg
30414137 10 g
30415358 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30416754 Supplier Toronto Research Chemicals Supplier No. TRCP22799050MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 63-92-3
Molecular Formula C18 H22 Cl N O . Cl H
Molecular Weight (g/mol) 340.29
Synonym Benzenemethanamine, N-(2-chloroethyl)-N-(1-methyl-2-phenoxyethyl)-, hydrochloride, Benzylamine, N-(2-chloroethyl)-N-(1-methyl-2-phenoxyethyl)-, hydrochloride, 2-(N-Benzyl-2-chloroethylamino)-1-phenoxypropane hydrochloride, Dibenzylin, Dibenzyline chloride, Dibenzyline hydrochloride, Dibenzyran, N-(2-Chloroethyl)-N-(1-methyl-2-phenoxyethyl)benzylamine, hydrochloride, N-2-Phenoxyisopropyl-N-benzyl-β-chloroethylamine hydrochloride, NSC 37448, Phenoxybenzamine chloride, Phenoxybenzamine hydrochloride
IUPAC Name N-benzyl-N-(2-chloroethyl)-1-phenoxypropan-2-amine;hydrochloride
SMILES Cl.CC(COc1ccccc1)N(CCCl)Cc2ccccc2
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C18H22ClNO.ClH/c1-16(15-21-18-10-6-3-7-11-18)20(13-12-19)14-17-8-4-2-5-9-17;/h2-11,16H,12-15H2,1H3;1H
Formula Weight 339.1157
Chemical Name or Material Phenoxybenzamine Hydrochloride