Learn More
Macherey-Nagel™ Phenolphthalein pH Paper
Product Code. 11306524
Click to view available options
Color Indicator:
White to Red
Unit Size:
1 roll
Product Code. 11306524
Brand: Macherey-Nagel™ 90712
Only null left
Add to basket
Add to basket
Macherey-Nagel Phenolphthalein pH Paper, Color Indicator: White to Red, pH Range: 8.3 to 10, Format: Reel, For Use With: Determination for neutralization analyses, Quantity: 1Each
Specifications
Color Indicator White to Red
pH Range 8.3 to 10
Format Reel
For Use With (Application) Determination for neutralization analyses
Quantity 1 Each
Type pH Paper