missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Glucuronide, Sodium Salt, TRC

Product Code. 30426898
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426898 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426898 Supplier Toronto Research Chemicals Supplier No. TRCP318018250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenolphthalein Glucuronide','Sodium Salt
CAS 6820-54-8
Molecular Formula C26H22O10 . Na
Quantity 250 mg
Synonym Phenolphthalein beta-D-glucuronide Sodium Salt, p-[3-(p-Hydroxyphenyl)phthalidyl]phenyl Glucosiduronic Acid Sodium Salt, 4-[1,3-Dihydro-1-(4-hydroxyphenyl)-3-oxo-1-isobenzofuranyl]phenyl beta-D-Glucopyranosiduronic Acid Monosodium Salt, p-[3-(p-Hydroxyphenyl)phthalidyl]phenyl beta-D-glucopyranosiduronic Acid Monosodium Salt
InChI Formula InChI=1S/C26H22O10.Na/c27-15-9-5-13(6-10-15)26(18-4-2-1-3-17(18)24(33)36-26)14-7-11-16(12-8-14)34-25-21(30)19(28)20(29)22(35-25)23(31)32;/h1-12,19-22,25,27-30H,(H,31,32);/q;+1/p-1/t19-,20-,21+,22-,25+,26?;/m0./s1
SMILES [Na+].O[C@H]1[C@H](Oc2ccc(cc2)C3(OC(=O)c4ccccc34)c5ccc(O)cc5)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)[O-]
IUPAC Name sodium;(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[4-[1-(4-hydroxyphenyl)-3-oxo-2-benzofuran-1-yl]phenoxy]oxane-2-carboxylate
Molecular Weight (g/mol) 516.4
Formula Weight 516.1032
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.