Learn More
DWK Life Sciences Kimble™ Phenolic Screw Cap with Liner
Click to view available options
Liner Material:
Rubber
Unit Size:
Pack of 1000
Description
Secure your media during autoclaving with the Kimble™ Phenolic Screw Cap with Liner. It is made of a special phenolic material (wood filled) and very resistant to the effects of temperature and steam encountered in autoclaving
- Styrene Liner
- Screw Cap
Specifications
Specifications
| Closure Size | 13-415 |
| Liner Material | Rubber |
| Autoclavable | Autoclavable |
| Description | Fixed Screw Cap |
| Material | Phenolic |
| Color | Black |
| Quantity | 1000 Pack |
| For Use With (Equipment) | Centrifuge Tubes |