Learn More
Phenol-OD, TRC
Specifications
Specifications
| Chemical Name or Material | Phenol-OD |
| CAS | 1003-66-3 |
| Molecular Formula | C6 D H5 O |
| Quantity | 50 mg |
| Synonym | Phenol-d (6CI','7CI','8CI','9CI','ACI), Phenol-d, Phenol-O-d, Phenol-O-D, Phenol-OD, (Hydroxy-d)benzene |
| InChI Formula | InChI=1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H/i/hD |
| SMILES | [2H]Oc1ccccc1 |
| IUPAC Name | deuteriooxybenzene |
| Molecular Weight (g/mol) | 95.117 |
| Formula Weight | 95.048 |