Learn More
Phenol-OD, CDN
Product Code. 30315108
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315108 5 g 5g
30319293 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315108 Supplier CDN Supplier No. CDND10585G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phenol-OD
CAS 1003-66-3
Molecular Formula C6 D H5 O
Quantity 5 g
Synonym Phenol-d (6 CI, 7 CI, 8 CI, 9 CI, ACI), Phenol-d, Phenol-O-d, Phenol-O-D, Phenol-OD, (Hydroxy-d)benzene
InChI Formula InChI=1 S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7 H/i/hD
SMILES [2 H]Oc1ccccc1
IUPAC Name deuteriooxybenzene
Molecular Weight (g/mol) 95.117
Formula Weight 95.048 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity