missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol-2,4,6-d3,OD, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | Phenol-2,4,6,O-D4 |
| CAS | 80441-87-8 |
| Molecular Formula | C6 D4 H2 O |
| Quantity | 0.1 g |
| Synonym | Phen-2,4,6-d3-ol-d (9 CI, ACI), 1,3,5-Trideuterio-2-deuteriooxybenzene, Phenol-2,4,6,O-D4 |
| InChI Formula | InChI=1 S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7 H/i1D,4 D,5 D/hD |
| SMILES | [2 H]Oc1c([2 H])cc([2 H])cc1[2 H] |
| IUPAC Name | 1,3,5-trideuterio-2-deuteriooxybenzene |
| Molecular Weight (g/mol) | 98.136 |
| Formula Weight | 98.067 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction