Learn More
Phe-Arg-Beta-naphthylamide Dihydrochloride, TRC
Product Code. 30413472
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30413472 10 mg
30419241 25 mg
30425909 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30413472 Supplier Toronto Research Chemicals Supplier No. TRCP99842510MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100929-99-5
Molecular Formula C25H32Cl2N6O2
Molecular Weight (g/mol) 519.47
Synonym L-Phenylalanyl-N-2-naphthalenyl-L-argininamide Dihydrochloride
IUPAC Name (S)-2-((S)-2-amino-3-phenylpropanamido)-5-guanidino-N-(naphthalen-2-yl)pentanamide dihydrochloride
SMILES N[C@H](C(N[C@@H](CCCNC(N)=N)C(NC1=CC=C(C=CC=C2)C2=C1)=O)=O)CC3=CC=CC=C3.Cl.Cl
Specifications
InChI Formula InChI=1S/C25H30N6O2.2ClH/c26-21(15-17-7-2-1-3-8-17)23(32)31-22(11-6-14-29-25(27)28)24(33)30-20-13-12-18-9-4-5-10-19(18)16-20;;/h1-5,7-10,12-13,16,21-22H,6,11,14-15,26H2,(H,30,33)(H,31,32)(H4,27,28,29);2*1H/t21-,22-;;/m0../s1
Quantity 10 mg
Formula Weight 518.2
Shipping Condition Room Temperature
Chemical Name or Material Phe-Arg-beta-naphthylamide Dihydrochloride