missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PHB2 Recombinant Protein (P02)

Human PHB2 full-length ORF recombinant protein with GST-tag at N-terminal

Brand:  Abnova™ H00011331-P02.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16166552

  • 508.00€ / 25µg
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

Sequence:
RESVFTVEGGHRAIFFNRIGGVQQDTILAEGLHFRIPWFQYPIIYDIRARPRKISSPTGSKDLQMVNISLRVLSRPNAQELPSMYQRLGLDYEERVLPSIVNEVLKSVVAKFNASQLITQRAQVSLLIRRELTERAKDFSLILDDVAITELSFSREYTAAVEAKQVAQQEAQRAQFLVEKAKQEQRQKIVQAEGEAEAAKMLGEALSKNPGYIKLRKIRAAQNISKTIATSQNRIYLTADNLVLNLQDESFTRGSDSLIKGKK
Best when used within three months from the date of receipt.ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications

Specifications

AAH14766
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
55
8
Glutathione Sepharose 4 Fast Flow
25 ug
RESVFTVEGGHRAIFFNRIGGVQQDTILAEGLHFRIPWFQYPIIYDIRARPRKISSPTGSKDLQMVNISLRVLSRPNAQELPSMYQRLGLDYEERVLPSIVNEVLKSVVAKFNASQLITQRAQVSLLIRRELTERAKDFSLILDDVAITELSFSREYTAAVEAKQVAQQEAQRAQFLVEKAKQEQRQKIVQAEGEAEAAKMLGEALSKNPGYIKLRKIRAAQNISKTIATSQNRIYLTADNLVLNLQDESFTRGSDSLIKGKK
BAP/BCAP37/Bap37/MGC117268/PNAS-141/REA/p22
PHB2
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
11331
PHB2 (Human) Recombinant Protein (P02)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PHB2
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers