Learn More
Cytiva PhastGel™ Buffer Strips
Product Code. p-3667695 Shop All Cytiva Products
Click to view available options
Quantity:
2 Pack
20 Strips
For Use With (Application):
Gel Electrophoresis
Product Type:
Buffer strip holder
PhastGel Buffer strips-SDS
PhastGel Buffer strips-native
Unit Size:
Pack of 10
Pack of 2
Pack of 20
Product Code. 10269294
Brand: Cytiva 18166801
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Pre-prepared 3% agarose blocks containing buffer, for use in conjunction with PhastGel gradient and homogeneous gel media to generate a multiphasic buffer system during electrophoresis
  • Ready made buffer strips eliminate the need to prepare electrophoresis buffers
  • Supplied as 10X pairs - for both cathode and anode
  • Choice of Native or Denaturing (SDS) buffer systems
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) PhastSystem Automated Electrophoresis Unit
Product Type Buffer strip holder
For Use With (Application) Gel Electrophoresis
Quantity 2 Pack