Learn More
Motic™ Phase Stop 0.5 Ph1 (10X, 20X, 40X)
Description
- Boost contrast and detail discrimination without staining or fixing, thus permitting live cell study.
- Must be matched and aligned to the objective phase plate.
Specifications
Specifications
| Type | Phase Stop Annulus |
| Description | 0.5 Ph1 (10X, 20X, 40X) |
| For Use With (Equipment) | AE31E Microscope, 10X, 20X, 40X Phase Objectives |