Learn More
Motic™ Phase Stop 0.3 Ph1 (10X, 20X, 40X)
Product Code. 15425362
Change view
Click to view available options
For Use With (Equipment):
AE31E Microscope, 10X, 20X, 40X Phase Objectives
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15425362 AE31E Microscope, 10X, 20X, 40X Phase Objectives Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15425362 Supplier Motic™ Supplier No. 1101000202391
Only null left
Add to basket
Add to basket
Use phase contrast to improve visualization of transparent specimens. Motic™ Phase Stop 0.3 Ph1 (10X, 20X, 40X) is an annular light stop that fits in your condenser.
  • Boost contrast and detail discrimination without staining or fixing, thus permitting live cell study.
  • Must be matched and aligned to the objective phase plate.
Specifications
Type Phase Stop Annulus
Description 0.3 Ph1 (10X, 20X, 40X)
For Use With (Equipment) AE31E Microscope, 10X, 20X, 40X Phase Objectives