Learn More
Olympus™ Phase Ring, IX-PH1U
Description
- Improve contrast.
- Improve visual detail.
- Fits IX microscopes.
Specifications
Specifications
| Description | Slit-ring style filter |
| For Use With (Application) | Increase visual contrast and visual detail via a phase-contrast approach |