Learn More
Olympus™ Phase Ring, IX-PH1U
Product Code. 12514077
Product Code. 12514077
Brand: Olympus™ K10017651
Increase visual contrast and visual detail via a phase-contrast approach. Olympus™ Phase Ring, IX-PH1U is a slit-ring style filter for IX microscopes.
  • Improve contrast.
  • Improve visual detail.
  • Fits IX microscopes.
Specifications
Description Slit-ring style filter
For Use With (Application) Increase visual contrast and visual detail via a phase-contrast approach