Learn More
Motic™ 5-Position Phase Contrast Turret Condenser
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
- 5 position turret condenser: BF, DF, Ph1, Ph2, Ph3
- For use with BA310/410 Microscsope
Order Info
dimProductType
Specifications
Specifications
| Type | Turret Condenser |
| For Use With (Application) | BF, DF, Ph1, Ph2, Ph3 phase contrast microscopy |
| Quantity | 1 Ea. |