Learn More
Cole-Parmer™ ThermoSafe™ Phase Change Soft Cold Packs
Product Code. 12338717 Shop All Cole-Parmer Products
Change view
Click to view available options
Dimensions (L x W x H):
17.1 x 10.2 x 2.5 cm
171 x 102 x 25 mm
191 x 165 x 25 mm
Unit Size:
Pack of 12
Pack of 24
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
12338717 191 x 165 x 25 mm Pack of 12
12328717 171 x 102 x 25 mm Pack of 24
12358717 17.1 x 10.2 x 2.5 cm Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12338717 Supplier Cole-Parmer™ Supplier No. 0634775
  • ONLINE EXCLUSIVE PRICE
    41.70€
    40.13€ / Pack of 12
    Save 1.57€
    3% Off
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
Use these cold packs for products requiring an accurate, consistent temperature. Cole-Parmer™ ThermoSafe™ Phase Soft Cold Packs are manufactured with laminated, metallized film exterior to resist punctures and provide a radiant heat barrier. Last fifteen percent longer than standard gel packs.
  • Available in 30 and °10°F (°1 and °23°C)  formulations.
  • High-tech phase change formula returns to gel state when thawed. 
  • Nontoxic and reusable
Specifications
Freezing Point -1°C
Dimensions (L x W x H) 191 x 165 x 25 mm