Learn More
Ismatec™ PharMed™ Ismaprene 3-stop Tubing
Product Code. p-7115966
Click to view available options
Tubing Size:
0.64mm I.D., 2.44mm O.D.
0.76m I.D., 2.36mm O.D.
1.30mm I.D., 2.80mm O.D.
2.79mm I.D., 4.59mm O.D.
Unit Size:
Pack of 12
Pack of 6
Product Code. 12201428
Brand: Ismatec™ SC0319A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ideal tubing for pharmaceutical and medical applications, including cell culture and tissue culture work, as well as foodstuffs. Ismatec™ PharMed™ Ismaprene 3-stop Tubing requires extension tubing on both suction and discharge sides.
  • Ideal for production filtration, fermentation and bioreactor process lines.
  • Very long service-life.
  • Non-toxic and non-hemolytic.
  • Impermeable to normal light and UV-radiation
  • Appropriate for medical products and foodstuffs.
  • Low particulate spallation.
  • Can be autoclaved repeatedly.
  • Withstands repeated CIP and SIP cleaning and sterilization.
  • Meets USP class VI, FDA and NSF criteria.
  • Potential leaching of additives (lubricants).
  • Applications
    • Acids: Good
    • Alkaline solutions: Good
    • Solvents: Not recommended
    • Pressure: Not recommended
    • Viscous media: Good
    • Sterile media: Excellent
  • Complies with: FDA 21CFR177.2600, US Pharmacopoea Class VI, NSF listed (Standard 51).
  • Sterilization
    • Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 141°C.
    • Gas sterilization with ethylene oxide.
    • Sterilization with radiation up to 2.5 mrad.
    • Caution: Use special tubing version (welded stoppers) when autoclaving 2 or 3-stop color-coded tubing.
Specifications
Certifications/Compliance FDA 21CFR177.2600 US Pharmacopoea Class VI NSF listed (Standard 51)
Description Color-code: Purple-White
Autoclavable Autoclavable
Material PharMed Ismaprene
Overall Length (Metric) 40 cm
Thickness (Metric) 0.90 mm
For Use With (Application) Pharmaceutical and medical applications, foodstuffs, cell cultures and tissue. MS/CA cassettes.
Min. Operating Temperature (Metric) -60°C
Packaging Quantity 6 pcs./Pk.
Tubing Size 2.79mm I.D., 4.59mm O.D.
Gas Permeability CO2 1200, O2 200, N2 80
Limitations Potential leaching of additives (lubricants).
Color Beige
Vacuum Level Excellent
Max. Operating Temperature (Metric) 135°C
Product Type 3-stop Tubing