Learn More
Masterflex™ PharMed™ BPT I/P™ High-Performance Precision Pump Tubing
Product Code. 11896683
Click to view available options
Tubing Size:
70
88
Unit Size:
Each
Product Code. 11896683
Brand: Masterflex™ 0650888
Only null left
Add to basket
Add to basket
Use this pump tubing for cell and tissue culture work and pharmaceutical applications. Masterflex™ PharMed™ BPT I/P™ High-Performance Precision Pump Tubing resists acids, alkalies, oxidizing agents, and animal and vegetable oils. Tubing is heat-sealable, bondable and formable. Good for light-sensitive samples.
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Always in stock — most formulations and sizes available to ship same day
  • Economical tubing with a longer tubing life
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner 12.7 mm
Material PharMed BPT
Overall Length (Metric) 762 cm
For Use With (Application) cell and tissue culture work and pharmaceutical uses
Min. Operating Temperature (Metric) -50°C
Packaging Quantity 1 Each
Resistance Properties Acids, Organic Solvents
Tubing Size 88
Flow Rate 0.02 to 17L/min.
Hose Barb 1/2in.
Height (Metric) 8.8 m
Color Opaque Beige
Maximum Pressure 35 psi
Vacuum Level 660 mm
Max. Operating Temperature (Metric) 74°C
Product Type Pump Tubing