Learn More
Esculape Laboratoire™ Pharmacy Cabinets
Product Code. p-8009249
Click to view available options
Product Type:
Pharmacy Cabinet - Empty
Pharmacy Cabinet - Filled
Height (Metric):
14 cm
20 cm
Unit Size:
Each
Product Code. 11738423
Brand: Esculape Laboratoire™ 711 03 04
Only null left
Specifications
Depth (Metric) 53 cm
For Use With (Application) To accommodate all the first aid equipment, offices, workshops and laboratories
Length (Metric) 53 cm
Product Type Pharmacy Cabinet - Filled
Quantity 1 Ea.
Height (Metric) 20 cm