Learn More
Sampling Systems™ Pharma Clear Labels
Shop All Sampling Systems Products
Click to view available options
Dimensions (LxW):
95 x 95 mm
Unit Size:
Pack of 500
Description
- Manufactured from a clear polypropylene film
- FDA-acceptable adhesive
- Ideal for sealing sampling holes
- Suitable for some types of NIR
- Each roll individually wrapped
- 95 x 95mm size
- Food-grade, medium-strength glue
- 500 labels per roll
Specifications
Specifications
| Certifications/Compliance | FDA food-safe |
| Color | Transparent |
| For Use With (Application) | Sealing sampling holes |
| Dimensions (LxW) | 95 x 95 mm |