Learn More
Proteintech Phantom Dye Red 780 Viability Dye
Phantom Dye Red 780 is a fixable viability dye which reacts with protein amine groups in both dead and live cells.
Brand:  Proteintech PD00002-100TESTS
Product Code. 17308252
  • 60.00€ / 100 tests
 Restricted Item
Estimated Shipment: 03-08-2026
Description
  • Due to the compromised cell membrane in dead cells the dye can bind to many more proteins inside the cell, resulting in much higher fluorescence and easy differentiation of Live/Dead cells.
  • Phantom Dye Red 780 is a water soluble red fluorescent dye excited by the red laser (633 nm) making it an excellent addition to your multicolor panel.