Learn More
Christeyns™ Phago'Derm Sensitive SPS Soap
Product Code. 17810265
Change view
Click to view available options
Volume (Metric):
500 mL
1000 mL
Quantity:
12 Airless Mounted Pump Bottles
12 Bottles
20 Square Mounted Pump Bottles
Unit Size:
12 L
Pack of 20
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Quantity Unit Size
17810265 1000 mL 12 Airless Mounted Pump Bottles 12 L
16618482 500 mL 20 Square Mounted Pump Bottles Pack of 20
17850245 1000 mL 12 Bottles 12 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17810265 Supplier Christeyns™ Supplier No. C65085
  • ONLINE EXCLUSIVE PRICE
    70.85€
    64.04€ / 12 L
    Save 6.81€
    9% Off
 Restricted Item
Estimated Shipment: 28-12-2026
Phago'Derm Sensitive SPS Soap is intended for simple washing of hands and forearms in intensive use. It preserves the protective barrier of the skin and provides a feeling of comfort.
  • Does not dry out the skin, even with frequent use
  • Hypoallergenic (formulated to minimize allergic reactions)
  • Suitable for washing children under 36 months and pregnant/breastfeeding women
  • Suitable for patient washing and preoperative showering
  • Good cleaning power.
  • High rinsability
  • Hydrating potential for 30 min.
Specifications
Product Type Hand Cleanser
Color Colorless
Form Foam
Quantity 12 Airless Mounted Pump Bottles
Volume (Metric) 1000 mL
compliance-icons
Product Identifier
  • 12LT X Soap PHAGODERM SENSITIVE SPS 1LT bottle Pum
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.