Learn More
Macherey-Nagel™ pH Paper
Product Code. 10613141
Click to view available options
pH Range:
0.5 to 5.5
1 to 11
1 to 14
3.8 to 5.8
4 to 7
5.4 to 7
5.5 to 9
6.4 to 8
7.2 to 9.7
9 to 13
Unit Size:
5m
Each
Pack of 5
Product Code. 10613141
Brand: Macherey-Nagel™ 90211
Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.
  • Universal and special indicator papers - simple pH measurement
  • Universal and special indicator papers supplied in plastic reels that ensure protection against external influences .
  • Always ready for use .
  • The reaction color of the paper is compared with the color scale and read the pH . Allows a very simple pH measurement .
Specifications
Certifications/Compliance ISO 9001
pH Range 7.2 to 9.7
Format Roll
Graduations <7.2, 7.2, 7.5, 7.8, 8.1, 8.4, 8.7, 9.0, 9.3, 9.7, >9.7
Quantity 1 Each
Type pH Paper