missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ pH/ORP/ISE pH Benchtop Meters

Product Code. 11872831
Change view
Click to view available options
Range (pH):
-2 to 20.00
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (pH) unitSize
11872831 -2 to 20.00 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11872831 Supplier Hanna Instruments™ Supplier No. HI322202

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Accuracy (ISE) ±0.5% of reading (monovalent ions), ±1% of reading (divalent ions)
Accuracy (mV) ±0.2 mV
Accuracy (pH) ±0.01/±0.002
Calibration Points Up to five point calibration, seven standard buffers available and two custom buffers
Calibration Points (ISE) Up to five point calibration, six standard solutions
Calibration Points (pH) Up to five point calibration, seven standard buffers available and two custom buffers
Detection Range –2 to 20.00°C
Display Type LCD
Range (mV) ±2000 mV
Range (pH) -2 to 20.00
Temperature Range (Metric) -20°C to +120.0°C
Inputs BNC
Model HI322202
Power Supply 230V
Product Type pH/ORP/ISE pH Benchtop Meter
Range (ISE) 1.00 E-7 to 9.99 E10 concentration
Resolution (mV) 0.1mV
Temperature Accuracy ±0.2°C
Temperature Compensation –20 to 120.0°C
Temperature (Metric) Operating -20°C to +120.0°C
Temperature Probe Calibration HI 7662-T temperature probe, stainless steel with 1 m (3.3’) cable (included)
Length (Metric) 235 mm
Height (Metric) 70 mm
Depth (Metric) 207 mm
Voltage 230 V
Frequency 50Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.