Learn More
Macherey-Nagel™ pH Neutral Litmus Paper
Click to view available options
pH Range:
5 to 8
Unit Size:
1 roll
Specifications
Specifications
| Color Indicator | Red to Violet to Blue |
| pH Range | 5 to 8 |
| Format | Reel |
| For Use With (Application) | Determination of acidic and alkaline reaction |
| Quantity | 1 Each |
| Type | pH Paper |
| Length (Metric) | 5000 mm |
| Width (Metric) | 7 mm |