Learn More
Macherey-Nagel™ pH Neutral Litmus Paper Strips
Click to view available options
pH Range:
8 to 5
Unit Size:
Pack of 100
Specifications
Specifications
| Color Indicator | Red to Violet to Blue |
| pH Range | 8 to 5 |
| Format | Strip |
| For Use With (Application) | Determination of acidic and alkaline reaction |
| Quantity | 100 Pack |
| Type | pH Paper |