Learn More
SI Analytics™ PH-METER Set HandyLab 680 Science
Product Code. 16379255 Shop All SI Analytics Products
Click to view available options
Range (pH):
0 to 14
Unit Size:
Each
Product Code. 16379255
Brand: SI Analytics™ 285204690
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The HandyLab 680 pH meter set with pHT-combination electrode A162 IDS and Z389 for demanding applications allows you to measure any two parameters simultaneously; pH, ORP, conductivity or oxygen.
  • Portable pH IDS meter
  • Two connectors for simultaneous measurement of pH, LF and DO
  • IDS technology
  • Auto-read function
  • CMC (Continuous measurement control)
  • QSC (Quality sensor control)
  • Traceability of the measuring values
  • User administration
  • Transmission of all data, in .csv format
  • Colour graphic display
  • Waterproof design IP67
TRUSTED_SUSTAINABILITY
Specifications
Data Management Details USB, USB Stick
Range (pH) 0 to 14
Calibration Points (pH) 1-, 2-, 3-, 4-, 5-points
Range (mV) ±1200.0, ±0.2
Temperature Range (Metric) -5°C to +105°C
Range (Conductivity) 0.00 to 2000 mS/cm ±0.5% of mean value
Range (Resistivity) 0.00 Ohm/cm to 100 Mohm/cm ±0.5% of mean value
Range (Salinity) 0.0 to 70.0 (IOT) ±0.5% of mean value
Range (TDS) 0 to 1999 mg/L, 0 to 199.9 g/L ±0.5% of mean value