Learn More
Macherey-Nagel™ pH Indicator Papers, Refill
Click to view available options
pH Range:
9 to 13
Unit Size:
Pack of 3
Description
- pH indicator papers have been on the market for decades and are the appreciated standard for many applications
- For each pH value these papers show a single color which can be matched with the color scale at intervals of 0.2–1 pH unit
Specifications
Specifications
| pH Range | 9 to 13 |
| Quantity | 1 x 5 m x 7 mm Spool |
| Type | pH Indicator Paper, Refill |
| Length (Metric) | 5 m |
| Width (Metric) | 7 mm |