Learn More
Crison™ 50 14 T pH Electrode
Description
- pH measuring range from 0 to 14
- Operating temperature range from −10° to +100°C
- Reference element of Ag/AgCl with AG+ ion barrier
- 2 ceramic diaphragms
- Min. immersion depth of 18mm
- CRISOLYT electrolyte
- Applications include aqueous samples, samples containing sulphide and reducing substances, samples with pH greater than 12, and punctual measurements up to 100°C
- Limitations include viscous solutions or solutions with colloids and distilled water
Specifications
Specifications
| For Use With (Application) | Measures aqueous media, suitable for samples containing sulphides, reducing sugars, or other substances which reacti with silver, and samples with pH >12 |
| Includes | Diaphragm: Ceramic, 2; Electrolyte: CRISOLYT, Pt1000 probe |
| Material | Glass |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | -10°C to +100°C |