Learn More
SI Analytics™ pH Combination Micro-Electrode
Product Code. 10223461 Shop All SI Analytics Products
Change view
Click to view available options
Connector Types:
BNC
DIN
DIN, 4 mm Banana Plug
Plug Head
Type:
Combination pH Micro Electrode
pH Combination Electrode
Membrane:
Cylinder
Spear
Sphere
Diameter (Metric):
12/3 mm
3 mm
5 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Type Membrane Diameter (Metric) Unit Size
10223461 BNC Combination pH Micro Electrode Cylinder 3 mm Each
10007900 DIN Combination pH Micro Electrode Cylinder 3 mm Each
10087870 DIN Combination pH Micro Electrode Spear 5 mm Each
10682551 DIN Combination pH Micro Electrode Sphere 5 mm Each
10251203 DIN, 4 mm Banana Plug pH Combination Electrode Sphere 12/3 mm Each
10467353 DIN pH Combination Electrode Cylinder 3 mm Each
10359761 Plug Head Combination pH Micro Electrode Cylinder 3 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10223461 Supplier SI Analytics™ Supplier No. 285105632
  • ONLINE EXCLUSIVE PRICE
    411.00€
    388.77€ / Pack of 1
    Save 22.23€
    5% Off
View all promotions
In Stock
Ensure high measurement accuracy, stability, and long service life while maintaining high adaptability to your measurement tasks. SI Analytics™ pH Combination Micro-Electrode is robust, durable, and easy to clean.
  • Proven high-end electrodes for demanding measurement tasks
  • Double junction Silamid™ reference system for fast and stable acquiring of measured values and for longer electrode life
  • Utmost versatility of pH electrodes is achieved by a large selection of junctions, membrane glass types and shapes, shaft lengths and diameters, ground joints, plug connections and integrated temperature sensors
  • Each pH combination electrode comes with individual serial number and quality certificate
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC
Junction Platinum Diaphragm
Material A-Glass
pH Range 7.0 ±0.3
Type Combination pH Micro Electrode
Internal Reference Ag/AgCl
Membrane Cylinder
Temperature (Metric) Range -5 to +100°C
Length (Metric) 96 mm
Length (Metric) Cable 1 m
Diameter (Metric) 3 mm