Learn More
Mettler Toledo™ pH Combination Electrodes
Product Code. 10425072 Shop All Mettler Toledo Products
Change view
Click to view available options
Length (Metric):
120 mm
pH Range:
0 to 12
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) pH Range Unit Size
10425072 120 mm 0 to 12 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10425072 Supplier Mettler Toledo™ Supplier No. 59903230
  • ONLINE EXCLUSIVE PRICE
    1320.60€
    521.74€ / Pack of 1
    Save 798.86€
    60% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Specifications
Type pH Combination Electrode
Length (Metric) 120 mm
pH Range 0 to 12
Temperature (Metric) Range 0°C to 100°C
For Use With (Equipment) Use in reactors, tanks, open basins and transfer lines under harsh chemical process conditions.