missing translation for 'onlineSavingsMsg'
Learn More

PFM 01

Product Code. 18707906
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18707906

Brand: Tocris Bioscience 6222/10

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

MRE11 endonuclease inhibitor

PFM 01 is a MRE11 endonuclease inhibitor. Prevents double-stranded break (DSB) resection. Enhances non-homologous end-joining (NHEJ) and reduces homologous recombination.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (5Z)-5-[(4-Hydroxyphenyl)methylene]-3-(2-methylpropyl)-2-thioxo-4-thiazolidinone
CAS 1558598-41-6
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C14H15NO2S2
Formula Weight Observed MW: 293.4
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.