Learn More
Cole-Parmer™ PFA Threaded Reducing Bushing
Product Code. 11720409
Click to view available options
Thread Size:
1/2 in. NPTM x 1/4 in. NPTF
1/2 in. NPTM x 3/8 in. NPTF
1/4 in. NPTM x 1/8 in. NPTF
3/4 in. NPTM x 1/2 in. NPTF
3/8 in. NPTM x 1/4 in. NPTF
3/8 in. NPTM x 1/8 in. NPTF
Unit Size:
Each
Product Code. 11720409
Brand: Cole-Parmer™ 0654608
Only null left
Add to basket
Add to basket
Threaded fittings offer more permanent installation
  • Chemically-inert PFA handles most aggressive chemicals
  • Threaded reducing bushing
  • Choose the size and material that meets your application needs
  • Use with our rigid tubing for your high-pressure applications
Specifications
Description NPT(M) x NPT(F) Connection
Autoclavable Autoclavable
Material PFA
Outlet Port 1/4 in.
Port Size 1/2 in.
Sterilization Method Autoclave or Ethylene Oxide
Thread Size 1/2 in. NPTM x 1/4 in. NPTF
Color Translucent
Product Type Threaded Reducing Bushing