Learn More
Masterflex™ Straight Adapter, Barb x NPT(M), PFA
Click to view available options
For Use With:
1/8 in. NPT(M) x 1/4 in. ID Tubing
Unit Size:
Each
Description
- PFA construction
- Easy to use
- Sterilizable via autoclave or ETO
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | PFA |
| For Use With | 1/8 in. NPT(M) x 1/4 in. ID Tubing |
| Packaging Quantity | 1 Pack |
| Tubing Size | Fits tubing accepting 1/8 in. NPT and 1/4 in. ID tubing. |
| Sterilization Method | Autoclave or Ethylene Oxide |
| Product Type | Threaded Adapter |