Learn More
Bohlender™ PFA Test Tube
Product Code. 10718252
Change view
Click to view available options
Capacity: 20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10718252 Capacity: 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10718252 Supplier Bohlender™ Supplier No. A18503
  • ONLINE EXCLUSIVE PRICE
    160.65€
    132.59€ / Pack of 1
    Save 28.06€
    17% Off
Estimated Shipment: 25-09-2026